Anti-ANGPT2 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB125692-100EA 840 EUR
ABCAAB125692-100
Anti-ANGPT2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ANGPT2 (angiopoietin 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR