Anti-IgG1/ IgG2b Rabbit Monoclonal Antibody

Lieferant: Abcam
ABCAAB126027-100EA 667 EUR
ABCAAB126027-100
Anti-IgG1/ IgG2b Rabbit Monoclonal Antibody
Antikörper
Type: Secondary
Antigen: IgG1/ IgG2b
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR