Anti-STK38L Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB126106-50EA 840 EUR
ABCAAB126106-50
Anti-STK38L Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: STK38L (serine/threonine kinase 38 like)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR