Anti-SGK3 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB126108-50EA 835 EUR
ABCAAB126108-50
Anti-SGK3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SGK3 (serum/glucocorticoid regulated kinase family, member 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR