Anti-RPL24 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB126172-50EA 835 EUR
ABCAAB126172-50
Anti-RPL24 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RPL24 (ribosomal protein L24)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR