Anti-TFAP2B Rabbit Polyclonal Antibody

Lieferant: Abcam
AB126207-50
ABCAAB126207-50EA 881 EUR
ABCAAB126207-50
Anti-TFAP2B Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TFAP2B (transcription factor AP-2 beta (activating enhancer binding protein 2 beta))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR