Anti-ENO3 Rabbit polyclonal antibody unconjugated

Lieferant: Abcam
ABCAAB126259-50EA 840 EUR
ABCAAB126259-50
Anti-ENO3 Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: ENO3 (enolase 3 (beta, muscle))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR