Anti-EXT1 Rabbit polyclonal antibody unconjugated
Lieferant: Abcam
ABCAAB126305-100EA 834 EUR
ABCAAB126305-100
Anti-EXT1 Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: EXT1 (exostosin glycosyltransferase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human