Anti-CCDC167 Rabbit Polyclonal Antibody

Lieferant: Abcam
AB126337-100
ABCAAB126337-100EA 857 EUR
ABCAAB126337-100
Anti-CCDC167 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CCDC167 (coiled-coil domain containing 167)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR