Anti-DPY30 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB126352-100EA 859 EUR
ABCAAB126352-100
Anti-DPY30 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: DPY30 (dpy-30 homolog (C. elegans))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR