Anti-LYRM5 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB126379-100EA 816 EUR
ABCAAB126379-100
Anti-LYRM5 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: LYRM5 (LYR motif containing 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR