Anti-FERD3L Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB126381-100EA 816 EUR
ABCAAB126381-100
Anti-FERD3L Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: FERD3L (Fer3-like bHLH transcription factor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR