Anti-TMEM97 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB126490-100EA 839 EUR
ABCAAB126490-100
Anti-TMEM97 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TMEM97 (transmembrane protein 97)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR