Anti-USP28 Rabbit Monoclonal Antibody [clone: EPR4249]
Lieferant: Abcam
ABCAAB126604-100EA 944 EUR
ABCAAB126604-100
Anti-USP28 Rabbit Monoclonal Antibody [clone: EPR4249]
Antikörper
Type: Primary
Antigen: USP28 (ubiquitin specific peptidase 28)
Clonality: monoclonal
Clone: EPR4249
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen