Anti-Sex Hormone Binding Globulin Rabbit Monoclonal Antibody [clone: EPR6628]
Lieferant: Abcam
ABCAAB126617-100EA 780 EUR
ABCAAB126617-100
Anti-Sex Hormone Binding Globulin Rabbit Monoclonal Antibody [clone: EPR6628]
Antikörper
Type: Primary
Antigen: Sex Hormone Binding Globulin
Clonality: monoclonal
Clone: EPR6628
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen