Anti-UCHL3 Rabbit Monoclonal Antibody [clone: EP5331]

Lieferant: Abcam
ABCAAB126621-100EA 782 EUR
ABCAAB126621-100
Anti-UCHL3 Rabbit Monoclonal Antibody [clone: EP5331]
Antikörper
Type: Primary
Antigen: UCHL3 (ubiquitin carboxyl-terminal esterase L3 (ubiquitin thiolesterase))
Clonality: monoclonal
Clone: EP5331
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR