Anti-Secretory Component Glycoprotein Rabbit Monoclonal Antibody [clone: EPR6701]

Lieferant: Abcam
ABCAAB126629-100EA 866 EUR
ABCAAB126629-100
Anti-Secretory Component Glycoprotein Rabbit Monoclonal Antibody [clone: EPR6701]
Antikörper
Type: Primary
Antigen: Secretory Component Glycoprotein
Clonality: monoclonal
Clone: EPR6701
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR