Anti-CARS Rabbit monoclonal antibody unconjugated [clone: EPR7121]

Lieferant: Abcam
ABCAAB126714-100EA 795 EUR
ABCAAB126714-100
Anti-CARS Rabbit monoclonal antibody unconjugated [clone: EPR7121]
Antikörper
Type: Primary
Antigen: CARS (cysteinyl-tRNA synthetase)
Clonality: monoclonal
Clone: EPR7121
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR