Anti-LMAN1 Rabbit Monoclonal Antibody [clone: EPR6980]

Lieferant: Abcam
AB126720-100
ABCAAB126720-100EA 896 EUR
ABCAAB126720-100
Anti-LMAN1 Rabbit Monoclonal Antibody [clone: EPR6980]
Antikörper
Type: Primary
Antigen: LMAN1 (lectin, mannose-binding, 1)
Clonality: monoclonal
Clone: EPR6980
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR