Anti-HLA-C Rabbit Monoclonal Antibody [clone: EPR6749]

Lieferant: Abcam
ABCAAB126722-100EA 785 EUR
ABCAAB126722-100
Anti-HLA-C Rabbit Monoclonal Antibody [clone: EPR6749]
Antikörper
Type: Primary
Antigen: HLA-C (major histocompatibility complex, class I, C)
Clonality: monoclonal
Clone: EPR6749
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR