Anti-DDX5 Rabbit monoclonal antibody unconjugated [clone: EPR7239]

Lieferant: Abcam
AB126730-100
ABCAAB126730-100EA 968 EUR
ABCAAB126730-100
Anti-DDX5 Rabbit monoclonal antibody unconjugated [clone: EPR7239]
Antikörper
Type: Primary
Antigen: DDX5 (DEAD (Asp-Glu-Ala-Asp) box helicase 5)
Clonality: monoclonal
Clone: EPR7239
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR