Anti-GAMT Rabbit monoclonal antibody unconjugated [clone: EPR6925]

Lieferant: Abcam
ABCAAB126736-100EA 777 EUR
ABCAAB126736-100
Anti-GAMT Rabbit monoclonal antibody unconjugated [clone: EPR6925]
Antikörper
Type: Primary
Antigen: GAMT (guanidinoacetate N-methyltransferase)
Clonality: monoclonal
Clone: EPR6925
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR