Anti-DMRT1 Rabbit Monoclonal Antibody [clone: EPR6936]

Lieferant: Abcam
ABCAAB126741-100EA 779 EUR
ABCAAB126741-100
Anti-DMRT1 Rabbit Monoclonal Antibody [clone: EPR6936]
Antikörper
Type: Primary
Antigen: DMRT1 (doublesex and mab-3 related transcription factor 1)
Clonality: monoclonal
Clone: EPR6936
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR