Anti-QK1 Rabbit Monoclonal Antibody [clone: EPR7306]

Lieferant: Abcam
AB126742-100
ABCAAB126742-100EA 885 EUR
ABCAAB126742-100
Anti-QK1 Rabbit Monoclonal Antibody [clone: EPR7306]
Antikörper
Type: Primary
Antigen: QK1
Clonality: monoclonal
Clone: EPR7306
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR