Anti-OGN Rabbit Monoclonal Antibody [clone: EPR6963]

Lieferant: Abcam
ABCAAB126749-100EA 778 EUR
ABCAAB126749-100
Anti-OGN Rabbit Monoclonal Antibody [clone: EPR6963]
Antikörper
Type: Primary
Antigen: OGN (osteoglycin)
Clonality: monoclonal
Clone: EPR6963
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR