Anti-CPSF2 Rabbit Monoclonal Antibody [clone: EPR7599]

Lieferant: Abcam
AB126760-100
ABCAAB126760-100EA 800 EUR
ABCAAB126760-100
Anti-CPSF2 Rabbit Monoclonal Antibody [clone: EPR7599]
Antikörper
Type: Primary
Antigen: CPSF2 (cleavage and polyadenylation specific factor 2, 100kDa)
Clonality: monoclonal
Clone: EPR7599
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR