Anti-THEMIS Rabbit Monoclonal Antibody [clone: EPR7353]

Lieferant: Abcam
ABCAAB126771-100EA 765 EUR
ABCAAB126771-100
Anti-THEMIS Rabbit Monoclonal Antibody [clone: EPR7353]
Antikörper
Type: Primary
Antigen: THEMIS (thymocyte selection associated)
Clonality: monoclonal
Clone: EPR7353
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR