Anti-SSR1 Rabbit Monoclonal Antibody [clone: EPR5604]

Lieferant: Abcam
AB126775-100
ABCAAB126775-100EA 816 EUR
ABCAAB126775-100
Anti-SSR1 Rabbit Monoclonal Antibody [clone: EPR5604]
Antikörper
Type: Primary
Antigen: SSR1 (signal sequence receptor, alpha)
Clonality: monoclonal
Clone: EPR5604
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR