Anti-PI 3 Kinase catalytic subunit alpha Mouse Monoclonal Antibody [clone: 4F3]

Lieferant: Abcam
ABCAAB126819-100EA 862 EUR
ABCAAB126819-100
Anti-PI 3 Kinase catalytic subunit alpha Mouse Monoclonal Antibody [clone: 4F3]
Antikörper
Type: Primary
Antigen: PI 3 Kinase catalytic subunit alpha
Clonality: monoclonal
Clone: 4F3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR