Anti-GPAA1 Rabbit Polyclonal Antibody

Lieferant: Abcam
AB126951-50
ABCAAB126951-50EA 837 EUR
ABCAAB126951-50
Anti-GPAA1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GPAA1 (glycosylphosphatidylinositol anchor attachment 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR