Anti-SNX7 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB126954-50EA 797 EUR
ABCAAB126954-50
Anti-SNX7 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SNX7 (sorting nexin 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR