Anti-PRMT7 Rabbit Polyclonal Antibody

Lieferant: Abcam
AB126965-50
ABCAAB126965-50EA 902 EUR
ABCAAB126965-50
Anti-PRMT7 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PRMT7 (protein arginine methyltransferase 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR