Anti-CES2 Rabbit polyclonal antibody unconjugated
Lieferant: Abcam
ABCAAB126970-50EA 840 EUR
ABCAAB126970-50
Anti-CES2 Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: CES2 (carboxylesterase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human