Anti-DCP2 Rabbit polyclonal antibody unconjugated
Lieferant: Abcam
ABCAAB127185-50EA 840 EUR
ABCAAB127185-50
Anti-DCP2 Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: DCP2 (decapping mRNA 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human