Anti-LARS Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB127754-50EA 840 EUR
ABCAAB127754-50
Anti-LARS Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: LARS (leucyl-tRNA synthetase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR