Anti-Helicobacter pylori urease B Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB127916-50EA 835 EUR
ABCAAB127916-50
Anti-Helicobacter pylori urease B Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Helicobacter pylori urease B
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR