Anti-FBN2 Rabbit polyclonal antibody unconjugated
Lieferant: Abcam
ABCAAB128026-50EA 835 EUR
ABCAAB128026-50
Anti-FBN2 Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: FBN2 (fibrillin 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human