Anti-NCKIPSD Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB128154-100EA 950 EUR
ABCAAB128154-100
Anti-NCKIPSD Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NCKIPSD (NCK interacting protein with SH3 domain)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR