Anti-SAMD1 Rabbit Polyclonal Antibody

Lieferant: Abcam
AB128170-100
ABCAAB128170-100EA 837 EUR
ABCAAB128170-100
Anti-SAMD1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SAMD1 (sterile alpha motif domain containing 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR