Anti-HHV6 Mouse Monoclonal Antibody [clone: C3108-103]

Lieferant: Abcam
ABCAAB128404-50EA 834 EUR
ABCAAB128404-50
Anti-HHV6 Mouse Monoclonal Antibody [clone: C3108-103]
Antikörper
Type: Primary
Antigen: HHV6
Clonality: monoclonal
Clone: C3108-103
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR