Anti-HSP90AA2 Mouse Monoclonal Antibody [clone: 5G5]
Lieferant: Abcam
ABCAAB128483-100EA 906 EUR
ABCAAB128483-100
Anti-HSP90AA2 Mouse Monoclonal Antibody [clone: 5G5]
Antikörper
Type: Primary
Antigen: HSP90AA2 (heat shock protein 90kDa alpha (cytosolic), class A member 2)
Clonality: monoclonal
Clone: 5G5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen