Anti-APLP2 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB128603-100EA 862 EUR
ABCAAB128603-100
Anti-APLP2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: APLP2 (amyloid beta (A4) precursor-like protein 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR