Anti-RSV glycoprotein Mouse Monoclonal Antibody [clone: 8C5 (9B6)]

Lieferant: Abcam
AB128708-200
ABCAAB128708-200EA 877 EUR
ABCAAB128708-200
Anti-RSV glycoprotein Mouse Monoclonal Antibody [clone: 8C5 (9B6)]
Antikörper
Type: Primary
Antigen: RSV glycoprotein
Clonality: monoclonal
Clone: 8C5 (9B6)
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR