Anti-IgA2 Mouse Monoclonal Antibody [clone: A9604D2] (Biotin)
Lieferant: Abcam
ABCAAB128731-250EA 834 EUR
ABCAAB128731-250
Anti-IgA2 Mouse Monoclonal Antibody [clone: A9604D2] (Biotin)
Antikörper
Type: Secondary
Antigen: IgA2
Clonality: monoclonal
Clone: A9604D2
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human