Anti-ATPB Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB128743-100EA 923 EUR
ABCAAB128743-100
Anti-ATPB Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ATPB
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR