Anti-IgA1 Mouse Monoclonal Antibody [clone: B3506B4]

Lieferant: Abcam
ABCAAB128791-250EA 835 EUR
ABCAAB128791-250
Anti-IgA1 Mouse Monoclonal Antibody [clone: B3506B4]
Antikörper
Type: Secondary
Antigen: IgA1
Clonality: Monoclonal
Clone: B3506B4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR