Anti-CTNNA3 Rabbit monoclonal antibody unconjugated [clone: EPR7601]

Lieferant: Abcam
ABCAAB128869-100EA 787 EUR
ABCAAB128869-100
Anti-CTNNA3 Rabbit monoclonal antibody unconjugated [clone: EPR7601]
Antikörper
Type: Primary
Antigen: CTNNA3 (catenin (cadherin-associated protein), alpha 3)
Clonality: monoclonal
Clone: EPR7601
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR