Anti-HTR7 Rabbit Monoclonal Antibody [clone: EPR6271]

Lieferant: Abcam
ABCAAB128892-100EA 834 EUR
ABCAAB128892-100
Anti-HTR7 Rabbit Monoclonal Antibody [clone: EPR6271]
Antikörper
Type: Primary
Antigen: HTR7 (5-hydroxytryptamine (serotonin) receptor 7, adenylate cyclase-coupled)
Clonality: monoclonal
Clone: EPR6271
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR