Anti-FOXO4 Rabbit monoclonal antibody unconjugated [clone: EPR5442]

Lieferant: Abcam
AB128908-100
ABCAAB128908-100EA 903 EUR
ABCAAB128908-100
Anti-FOXO4 Rabbit monoclonal antibody unconjugated [clone: EPR5442]
Antikörper
Type: Primary
Antigen: FOXO4 (forkhead box O4)
Clonality: monoclonal
Clone: EPR5442
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR