Anti-ACAA2 Rabbit Monoclonal Antibody [clone: EPR6733]

Lieferant: Abcam
AB128911-100
ABCAAB128911-100EA 818 EUR
ABCAAB128911-100
Anti-ACAA2 Rabbit Monoclonal Antibody [clone: EPR6733]
Antikörper
Type: Primary
Antigen: ACAA2 (acetyl-CoA acyltransferase 2)
Clonality: monoclonal
Clone: EPR6733
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR